RIE240870366 Rieke Metals, Inc.

4-(4-Trifluoromethylbenzoyl)isoquinoline

  • MDL number MFCD13153017
  • Molecular Weight : 301.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-10
Availability details for
Quantity
  • Availability
  • Add to Cart