RIE240870381 Rieke Metals, Inc.

4-(3-Phenoxybenzoyl)isoquinoline

  • MDL number MFCD13153032
  • Molecular Weight : 325.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isoquinolinyl(3-phenoxyphenyl)methanone
Inchi Code 1S/C22H15NO2/c24-22(21-15-23-14-17-7-4-5-12-20(17)21)16-8-6-11-19(13-16)25-18-9-2-1-3-10-18/h1-15H
Inchi Key CITPUHZGDNAPNG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-27
Availability details for
Quantity
  • Availability
  • Add to Cart