RIE240870382 Rieke Metals, Inc.

4-(4-Phenoxybenzoyl)isoquinoline

  • MDL number MFCD13153033
  • Molecular Weight : 325.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isoquinolinyl(4-phenoxyphenyl)methanone
Inchi Code 1S/C22H15NO2/c24-22(21-15-23-14-17-6-4-5-9-20(17)21)16-10-12-19(13-11-16)25-18-7-2-1-3-8-18/h1-15H
Inchi Key URKGCNMYMMFQEG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-28
Availability details for
Quantity
  • Availability
  • Add to Cart