RIE240870384 Rieke Metals, Inc.

4-(4-Nitrobenzoyl)isoquinoline

  • MDL number MFCD13153035
  • Molecular Weight : 278.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isoquinolinyl(4-nitrophenyl)methanone
Inchi Code 1S/C16H10N2O3/c19-16(11-5-7-13(8-6-11)18(20)21)15-10-17-9-12-3-1-2-4-14(12)15/h1-10H
Inchi Key FSXLAFAJNPKRIU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-31
Availability details for
Quantity
  • Availability
  • Add to Cart