RIE240870386 Rieke Metals, Inc.

4-(4-Isopropoxybenzoyl)isoquinoline

  • MDL number MFCD13153037
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-33
Availability details for
Quantity
  • Availability
  • Add to Cart