RIE240870387 Rieke Metals, Inc.

4-(4-tert-Butylbenzoyl)isoquinoline

  • MDL number MFCD13153038
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-34
Availability details for
Quantity
  • Availability
  • Add to Cart