RIE240870390 Rieke Metals, Inc.

4-(4-Butylbenzoyl)isoquinoline

  • MDL number MFCD13153041
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butylphenyl)(4-isoquinolinyl)methanone
Inchi Code 1S/C20H19NO/c1-2-3-6-15-9-11-16(12-10-15)20(22)19-14-21-13-17-7-4-5-8-18(17)19/h4-5,7-14H,2-3,6H2,1H3
Inchi Key DZCOUWJTZGSCPV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-35c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-35c
Availability details for
Quantity
  • Availability
  • Add to Cart