RIE240870397 Rieke Metals, Inc.

4-(4-Pentyloxybenzoyl)isoquinoline

  • MDL number MFCD13153048
  • Molecular Weight : 319.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-36D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-36D
Availability details for
Quantity
  • Availability
  • Add to Cart