RIE240870403 Rieke Metals, Inc.

4-(2,6-Difluorobenzoyl)isoquinoline

  • MDL number MFCD13153054
  • Molecular Weight : 269.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-difluorophenyl)(4-isoquinolinyl)methanone
Inchi Code 1S/C16H9F2NO/c17-13-6-3-7-14(18)15(13)16(20)12-9-19-8-10-4-1-2-5-11(10)12/h1-9H
Inchi Key JFAICDLYINSBRY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-40

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-40
Availability details for
Quantity
  • Availability
  • Add to Cart