RIE240870413 Rieke Metals, Inc.

4-(2,4-Dimethoxybenzoyl)isoquinoline

  • MDL number MFCD13153064
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-dimethoxyphenyl)(4-isoquinolinyl)methanone
Inchi Code 1S/C18H15NO3/c1-21-13-7-8-15(17(9-13)22-2)18(20)16-11-19-10-12-5-3-4-6-14(12)16/h3-11H,1-2H3
Inchi Key AEVOGXUGUFHOFZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-50
Availability details for
Quantity
  • Availability
  • Add to Cart