RIE240870419 Rieke Metals, Inc.

4-(2,4-Dimethylbenzoyl)isoquinoline

  • MDL number MFCD13153070
  • Molecular Weight : 261.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-56
Availability details for
Quantity
  • Availability
  • Add to Cart