RIE240870423 Rieke Metals, Inc.

4-(3,5-Dimethylbenzoyl)isoquinoline

  • MDL number MFCD13153074
  • Molecular Weight : 261.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dimethylphenyl)(4-isoquinolinyl)methanone
Inchi Code 1S/C18H15NO/c1-12-7-13(2)9-15(8-12)18(20)17-11-19-10-14-5-3-4-6-16(14)17/h3-11H,1-2H3
Inchi Key OJNZLVDSTFJJEX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-60
Availability details for
Quantity
  • Availability
  • Add to Cart