RIE240870426 Rieke Metals, Inc.

4-(2-Trifluoromethylbenzoyl)quinoline

  • MDL number MFCD13153080
  • Molecular Weight : 301.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-08
Availability details for
Quantity
  • Availability
  • Add to Cart