RIE240870434 Rieke Metals, Inc.

3-(4-Iodobenzoyl)quinoline

  • MDL number MFCD13153088
  • Molecular Weight : 359.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-iodophenyl)(3-quinolinyl)methanone
Inchi Code 1S/C16H10INO/c17-14-7-5-11(6-8-14)16(19)13-9-12-3-1-2-4-15(12)18-10-13/h1-10H
Inchi Key MOLSQANUBBVFDQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-22
Availability details for
Quantity
  • Availability
  • Add to Cart