RIE240870436 Rieke Metals, Inc.

3-(4-Cyanobenzoyl)quinoline

  • MDL number MFCD13153090
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-quinolinylcarbonyl)benzonitrile
Inchi Code 1S/C17H10N2O/c18-10-12-5-7-13(8-6-12)17(20)15-9-14-3-1-2-4-16(14)19-11-15/h1-9,11H
Inchi Key SVSNMICTLGLMEM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-25
Availability details for
Quantity
  • Availability
  • Add to Cart