RIE240870442 Rieke Metals, Inc.

3-(4-tert-butylbenzoyl)quinoline

  • MDL number MFCD13153097
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)(3-quinolinyl)methanone
Inchi Code 1S/C20H19NO/c1-20(2,3)17-10-8-14(9-11-17)19(22)16-12-15-6-4-5-7-18(15)21-13-16/h4-13H,1-3H3
Inchi Key AZUHVXQGYCYEMD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-34
Availability details for
Quantity
  • Availability
  • Add to Cart