RIE240870447 Rieke Metals, Inc.

4-(4-Heptylbenzoyl)quinoline

  • MDL number MFCD13153103
  • Molecular Weight : 331.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-35F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-35F
Availability details for
Quantity
  • Availability
  • Add to Cart