RIE240870450 Rieke Metals, Inc.

4-(4-Butoxybenzoyl)quinoline

  • MDL number MFCD13153106
  • Molecular Weight : 305.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-36C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-36C
Availability details for
Quantity
  • Availability
  • Add to Cart