RIE240870455 Rieke Metals, Inc.

4-(2,4-Difluorobenzoyl)quinoline

  • MDL number MFCD13153111
  • Molecular Weight : 269.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-38

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-38
Availability details for
Quantity
  • Availability
  • Add to Cart