RIE240870458 Rieke Metals, Inc.

4-(3,5-Difluorobenzoyl)quinoline

  • MDL number MFCD13153115
  • Molecular Weight : 269.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-42
Availability details for
Quantity
  • Availability
  • Add to Cart