RIE240870459 Rieke Metals, Inc.

3-(2,3-Dichlorobenzoyl)quinoline

  • MDL number MFCD13153116
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dichlorophenyl)(3-quinolinyl)methanone
Inchi Code 1S/C16H9Cl2NO/c17-13-6-3-5-12(15(13)18)16(20)11-8-10-4-1-2-7-14(10)19-9-11/h1-9H
Inchi Key KRXMTUNVDLPGPZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-43
Availability details for
Quantity
  • Availability
  • Add to Cart