RIE240870460 Rieke Metals, Inc.

4-(2,4-Dichlorobenzoyl)quinoline

  • MDL number MFCD13153117
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-44
Availability details for
Quantity
  • Availability
  • Add to Cart