RIE240870464 Rieke Metals, Inc.

4-(3,5-Dichlorobenzoyl)quinoline

  • MDL number MFCD13153121
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-48
Availability details for
Quantity
  • Availability
  • Add to Cart