RIE240870468 Rieke Metals, Inc.

3-(2,6-Dimethoxybenzoyl)quinoline

  • MDL number MFCD13153125
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-dimethoxyphenyl)(3-quinolinyl)methanone
Inchi Code 1S/C18H15NO3/c1-21-15-8-5-9-16(22-2)17(15)18(20)13-10-12-6-3-4-7-14(12)19-11-13/h3-11H,1-2H3
Inchi Key JYNYZXXRUDORLK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-52
Availability details for
Quantity
  • Availability
  • Add to Cart