RIE240870470 Rieke Metals, Inc.

4-(3,5-Dimethoxybenzoyl)quinoline

  • MDL number MFCD13153127
  • Molecular Weight : 293.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-54
Availability details for
Quantity
  • Availability
  • Add to Cart