RIE240870474 Rieke Metals, Inc.

4-(2,6-Dimethylbenzoyl)quinoline

  • MDL number MFCD13153131
  • Molecular Weight : 261.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-58
Availability details for
Quantity
  • Availability
  • Add to Cart