RIE240870477 Rieke Metals, Inc.

4-(3,4,5-Trifluorobenzoyl)quinoline

  • MDL number MFCD13153134
  • Molecular Weight : 287.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-61
Availability details for
Quantity
  • Availability
  • Add to Cart