RIE240870478 Rieke Metals, Inc.

4-(Pentafluorobenzoyl)quinoline

  • MDL number MFCD13153135
  • Molecular Weight : 323.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-62
Availability details for
Quantity
  • Availability
  • Add to Cart