RIE240870481 Rieke Metals, Inc.

3-(3-Methoxybenzoyl)-4-methylpyridine

  • MDL number MFCD13153138
  • Molecular Weight : 227.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5622-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-03
Availability details for
Quantity
  • Availability
  • Add to Cart