RIE240870484 Rieke Metals, Inc.

3-(3-Methylbenzoyl)-4-methylpyridine

  • MDL number MFCD13153141
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methylphenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C14H13NO/c1-10-4-3-5-12(8-10)14(16)13-9-15-7-6-11(13)2/h3-9H,1-2H3
Inchi Key KJMXXHJCGXQFJQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-06
Availability details for
Quantity
  • Availability
  • Add to Cart