RIE240870485 Rieke Metals, Inc.

3-(4-Methylbenzoyl)-4-methylpyridine

  • MDL number MFCD13153142
  • Molecular Weight : 211.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5622-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-07
Availability details for
Quantity
  • Availability
  • Add to Cart