RIE240870489 Rieke Metals, Inc.

3-(2-Fluorobenzoyl)-4-methylpyridine

  • MDL number MFCD13153145
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-fluorophenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C13H10FNO/c1-9-6-7-15-8-11(9)13(16)10-4-2-3-5-12(10)14/h2-8H,1H3
Inchi Key KQQLLCUYCVXRBJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-11
Availability details for
Quantity
  • Availability
  • Add to Cart