RIE240870493 Rieke Metals, Inc.

3-(3-Chlorobenzoyl)-4-methylpyridine

  • MDL number MFCD13153149
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5622-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-15
Availability details for
Quantity
  • Availability
  • Add to Cart