RIE240870496 Rieke Metals, Inc.

3-(3-Bromobenzoyl)-4-methylpyridine

  • MDL number MFCD13153152
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C13H10BrNO/c1-9-5-6-15-8-12(9)13(16)10-3-2-4-11(14)7-10/h2-8H,1H3
Inchi Key KUKOGUNUOZCEBL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-18
Availability details for
Quantity
  • Availability
  • Add to Cart