RIE240870511 Rieke Metals, Inc.

4-Methyl-3-(4-propylbenzoyl)pyridine

  • MDL number MFCD13153167
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5622-35B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-35B
Availability details for
Quantity
  • Availability
  • Add to Cart