RIE240870513 Rieke Metals, Inc.

4-Methyl-3-(4-pentylbenzoyl)pyridine

  • MDL number MFCD13153169
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methyl-3-pyridinyl)(4-pentylphenyl)methanone
Inchi Code 1S/C18H21NO/c1-3-4-5-6-15-7-9-16(10-8-15)18(20)17-13-19-12-11-14(17)2/h7-13H,3-6H2,1-2H3
Inchi Key JLYRBPJHZDZUKI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-35d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-35d
Availability details for
Quantity
  • Availability
  • Add to Cart