RIE240870514 Rieke Metals, Inc.

3-(4-Hexylbenzoyl)-4-methylpyridine

  • MDL number MFCD13153170
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-hexylphenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C19H23NO/c1-3-4-5-6-7-16-8-10-17(11-9-16)19(21)18-14-20-13-12-15(18)2/h8-14H,3-7H2,1-2H3
Inchi Key LRDGLRVZLJJHJG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-35e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-35e
Availability details for
Quantity
  • Availability
  • Add to Cart