RIE240870515 Rieke Metals, Inc.

3-(4-Heptylbenzoyl)-4-methylpyridine

  • MDL number MFCD13153171
  • Molecular Weight : 295.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-heptylphenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C20H25NO/c1-3-4-5-6-7-8-17-9-11-18(12-10-17)20(22)19-15-21-14-13-16(19)2/h9-15H,3-8H2,1-2H3
Inchi Key MQDNZJBZBXKVMG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-35f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-35f
Availability details for
Quantity
  • Availability
  • Add to Cart