RIE240870520 Rieke Metals, Inc.

3-(4-Hexyloxybenzoyl)-4-methylpyridine

  • MDL number MFCD13153176
  • Molecular Weight : 297.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(hexyloxy)phenyl](4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C19H23NO2/c1-3-4-5-6-13-22-17-9-7-16(8-10-17)19(21)18-14-20-12-11-15(18)2/h7-12,14H,3-6,13H2,1-2H3
Inchi Key RRESOPFBRLCHDY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-36e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-36e
Availability details for
Quantity
  • Availability
  • Add to Cart