RIE240870522 Rieke Metals, Inc.

3-(2,3-Difluorobenzoyl)-4-methylpyridine

  • MDL number MFCD13153178
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-difluorophenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-5-6-16-7-10(8)13(17)9-3-2-4-11(14)12(9)15/h2-7H,1H3
Inchi Key HTNTXOLWFUWHPW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-37
Availability details for
Quantity
  • Availability
  • Add to Cart