RIE240870527 Rieke Metals, Inc.

3-(3,5-Difluorobenzoyl)-4-methylpyridine

  • MDL number MFCD13153183
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-2-3-16-7-12(8)13(17)9-4-10(14)6-11(15)5-9/h2-7H,1H3
Inchi Key KYEFRHOYPYOVNS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-42
Availability details for
Quantity
  • Availability
  • Add to Cart