RIE240870535 Rieke Metals, Inc.

3-(2,4-Dimethoxybenzoyl)-4-methylpyridine

  • MDL number MFCD13153191
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-dimethoxyphenyl)(4-methyl-3-pyridinyl)methanone
Inchi Code 1S/C15H15NO3/c1-10-6-7-16-9-13(10)15(17)12-5-4-11(18-2)8-14(12)19-3/h4-9H,1-3H3
Inchi Key IAYNUPHRUKWZJS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-50
Availability details for
Quantity
  • Availability
  • Add to Cart