RIE240870545 Rieke Metals, Inc.

3-(3,5-Dimethylbenzoyl)-4-methylpyridine

  • MDL number MFCD13153201
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5622-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-60
Availability details for
Quantity
  • Availability
  • Add to Cart