RIE240870546 Rieke Metals, Inc.

4-Methyl-3-(3,4,5-trifluorobenzoyl)pyridine

  • MDL number MFCD13153202
  • Molecular Weight : 251.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methyl-3-pyridinyl)(3,4,5-trifluorophenyl)methanone
Inchi Code 1S/C13H8F3NO/c1-7-2-3-17-6-9(7)13(18)8-4-10(14)12(16)11(15)5-8/h2-6H,1H3
Inchi Key XHRVTPTYCAIDEP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-61
Availability details for
Quantity
  • Availability
  • Add to Cart