RIE240870547 Rieke Metals, Inc.

4-Methyl-3-(pentafluorobenzoyl)pyridine

  • MDL number MFCD13153203
  • Molecular Weight : 287.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methyl-3-pyridinyl)(2,3,4,5,6-pentafluorophenyl)methanone
Inchi Code 1S/C13H6F5NO/c1-5-2-3-19-4-6(5)13(20)7-8(14)10(16)12(18)11(17)9(7)15/h2-4H,1H3
Inchi Key UWOLUKAINNTPKK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5622-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5622-62
Availability details for
Quantity
  • Availability
  • Add to Cart