RIE240870548 Rieke Metals, Inc.

5-Benzoyl-2-methylpyridine

  • MDL number MFCD08272280
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-01
Availability details for
Quantity
  • Availability
  • Add to Cart