RIE240870550 Rieke Metals, Inc.

5-(3-Methoxybenzoyl)-2-methylpyridine

  • MDL number MFCD13153205
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxyphenyl)(6-methyl-3-pyridinyl)methanone
Inchi Code 1S/C14H13NO2/c1-10-6-7-12(9-15-10)14(16)11-4-3-5-13(8-11)17-2/h3-9H,1-2H3
Inchi Key LNYGTQKUVZJCDE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5623-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-03
Availability details for
Quantity
  • Availability
  • Add to Cart