RIE240870563 Rieke Metals, Inc.

5-(4-Chlorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153218
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-16
Availability details for
Quantity
  • Availability
  • Add to Cart