RIE240870565 Rieke Metals, Inc.

5-(3-Bromobenzoyl)-2-methylpyridine

  • MDL number MFCD13153220
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-18
Availability details for
Quantity
  • Availability
  • Add to Cart