RIE240870570 Rieke Metals, Inc.

5-(3-Cyanobenzoyl)-2-methylpyridine

  • MDL number MFCD13153225
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-24

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-24
Availability details for
Quantity
  • Availability
  • Add to Cart